Order Entry
Germany
ContactUsLinkComponent
1-[Amino(4-nitrophenyl)methyl]naphthalen-2-ol hydrochloride
1-[Amino(4-nitrophenyl)methyl]naphthalen-2-ol hydrochloride
Catalog # APOSOR307609-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[Amino(4-nitrophenyl)methyl]naphthalen-2-ol hydrochloride
Catalog # APOSOR307609-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Storage Temperature:Ambient
CAS Number:714953-84-1
Formula:C₁₇H₁₅ClN₂O₃
MDL Number:MFCD07366591