tert-Butyl-5-amino-1H-indazole-1-carboxylate
Catalog # APOSOR30732-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:233.27 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:129488-10-4
Formula:C₁₂H₁₅N₃O₂
MDL Number:MFCD04114656