Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl-3-bromo-1H-pyrazole-4-carboxylate
Ethyl-3-bromo-1H-pyrazole-4-carboxylate
Ethyl-3-bromo-1H-pyrazole-4-carboxylate
Irritant
Ethyl-3-bromo-1H-pyrazole-4-carboxylate
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Ethyl-3-bromo-1H-pyrazole-4-carboxylate
Supplier:
Apollo Scientific
About this item
CAS Number:
1353100-91-0
:
Formula:
C₆H₇BrN₂O₂
MDL Number:
MFCD12032195
:
219.04 g/mol
Melting Pt:
85…88 °C
:
GHS07
Storage Temperature:
Ambient