
Methyl-1H-indazole-5-carboxylate
Catalog # APOSOR30715-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:176.1700 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:473416-12-5
Formula:C₉H₈N₂O₂
MDL Number:MFCD07371609