Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl-1H-imidazole-5-carboxylate
Methyl-1H-imidazole-5-carboxylate 98%
Methyl-1H-imidazole-5-carboxylate 98%
:
Apollo Scientific
Methyl-1H-imidazole-5-carboxylate 98%
:
Apollo Scientific
:
17325-26-7
:
:
C₅H₆N₂O₂
:
MFCD00216584
:
126.11 g/mol
:
154…156 °C
:
GHS07
:
Ambient