Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-1H-pyrrolo[3,2-b]pyridine
3-Bromo-1H-pyrrolo[3,2-b]pyridine 95%
3-Bromo-1H-pyrrolo[3,2-b]pyridine 95%
:
Apollo Scientific
3-Bromo-1H-pyrrolo[3,2-b]pyridine 95%
:
Apollo Scientific
:
23688-47-3
:
:
C₇H₅BrN₂
:
MFCD10697525
:
197.04 g/mol
:
GHS07
:
Ambient