6-Amino-2,3-dihydroindole-1-carboxylic acid tert-butyl ester
Catalog # APOSOR305255-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:234.3 g/mol
Storage Temperature:Ambient
CAS Number:129488-00-2
Formula:C₁₃H₁₈N₂O₂
MDL Number:MFCD08059271