(R)-(-)-2-Butanol 99+%
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: (R)-(-)-Ethyl methyl carbinol, (R)-(-)-sec-Butyl alcohol, (R)-(-)-2-Hydroxy butane, (R)-(-)-sec-Butanol, (R)-(-)-Methyl ethyl carbinol
- CAS Number:14898-79-4
- Formula:C₄H₁₀O
- MDL Number:MFCD00064280
- Molecular Weight:74.12 g/mol
- Boiling Pt:99…100 °C
- Melting Pt:–114 °C
- Density:0.806 g/cm³ (25 °C)
- Flash Pt:26 °C
- Storage Temperature:Ambient
- UN:1120
- ADR:3,II
3 Options Available Below
Product Details & Documents
Synonyms: (R)-(-)-Ethyl methyl carbinol, (R)-(-)-sec-Butyl alcohol, (R)-(-)-2-Hydroxy butane, (R)-(-)-sec-Butanol, (R)-(-)-Methyl ethyl carbinol
- CAS Number:14898-79-4
- Formula:C₄H₁₀O
- MDL Number:MFCD00064280
- Molecular Weight:74.12 g/mol
- Boiling Pt:99…100 °C
- Melting Pt:–114 °C
- Density:0.806 g/cm³ (25 °C)
- Flash Pt:26 °C
- Storage Temperature:Ambient
- UN:1120
- ADR:3,II



