1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazole-4-carboxylic acid 97%
Catalog # APOSOR303023-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:232.24 g/mol
Storage Temperature:Ambient
CAS Number:83-10-3
Formula:C₁₂H₁₂N₂O₃
MDL Number:MFCD00020785