2-Hydroxy-5-nitropyridine
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 5-Nitro-2-pyridinol
- CAS Number:5418-51-9
- Formula:C₅H₄N₂O₃
- MDL Number:MFCD00006276
- Molecular Weight:140.1 g/mol
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
Synonyms: 5-Nitro-2-pyridinol
- CAS Number:5418-51-9
- Formula:C₅H₄N₂O₃
- MDL Number:MFCD00006276
- Molecular Weight:140.1 g/mol
- Storage Temperature:Ambient



