
3,5-Dimethylisoxazole 95%
Catalog # APOSOR30211-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:97.12 g/mol
Boiling Pt:142…144 °C
Density:0.99 g/cm³ (25 °C)
Flash Pt:31 °C
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS02
CAS Number:300-87-8
Formula:C₅H₇NO
MDL Number:MFCD00003156