Order Entry
Germany
ContactUsLinkComponent
3-Methylbenzyl mercaptan 95%
Irritant
3-Methylbenzyl mercaptan 95%
Catalog # APOSOR30177-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Methylbenzyl mercaptan 95%
Catalog # APOSOR30177-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :138.23
Boiling Pt:102…104 °C/15mmHg
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
 :GHS07
CAS Number:25697-56-7
Formula:C₈H₁₀S
MDL Number:MFCD00022075