Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4,5-Tribromotoluene
2,4,5-Tribromotoluene
2,4,5-Tribromotoluene
APOSOR30148-25G
:
Apollo Scientific
:
2,4,5-Tribromotoluene
APOSOR30148-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
112…115 °C
:
328.83 g/mol
:
GHS07
:
Ambient
:
3278-88-4
:
C₇H₅Br₃
:
MFCD00052984