Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
tert-Butyl-3-oxoazepane-1-carboxylate
tert-Butyl-3-oxoazepane-1-carboxylate
tert-Butyl-3-oxoazepane-1-carboxylate
tert-Butyl-3-oxoazepane-1-carboxylate
APOSOR301179-1G
:
Apollo Scientific
:
tert-Butyl-3-oxoazepane-1-carboxylate
APOSOR301179-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
90 °C/0.15mmHg
:
213.2735 g/mol
:
GHS07
:
Refrigerator
:
870842-23-2
:
C₁₁H₁₉NO₃
:
MFCD08443803