
Methyl-3-aminobenzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:4518-10-9
:
Formula:C₈H₉NO₂
MDL Number:MFCD00017102
:151.16 g/mol
Boiling Pt:170 °C/20mmHg
Melting Pt:49…55 °C
Storage Temperature:Refrigerator
EINECS:224-842-5
:GHS07