tert-Butyl-2-formylpiperidine-1-carboxylate 97%
Catalog # APOSOR300652-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:213.2700 g/mol
Storage Temperature:Ambient
CAS Number:157634-02-1
Formula:C₁₁H₁₉NO₃
MDL Number:MFCD02179021