Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl-1H-pyrrole-2-carboxylate
Methyl-1H-pyrrole-2-carboxylate
Methyl-1H-pyrrole-2-carboxylate
Irritant
Methyl-1H-pyrrole-2-carboxylate
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Methyl-1H-pyrrole-2-carboxylate
Supplier:
Apollo Scientific
About this item
CAS Number:
1193-62-0
:
Formula:
C₆H₇NO₂
MDL Number:
MFCD00817048
:
125.13 g/mol
Melting Pt:
74…78 °C
:
GHS07
Storage Temperature:
Ambient