7-Isopropyl-1,4-dimethylazulene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:489-84-9
:
Formula:C₁₅H₁₈
MDL Number:MFCD00003811
:198.31 g/mol
Boiling Pt:153 °C/7mmHg
Melting Pt:27 °C
Density:0.976 g/cm³ (25 °C)
Flash Pt:113 °C
:GHS07
Storage Temperature:Ambient