Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Methyl-1-p-tolylethylamine
1-Methyl-1-p-tolylethylamine
1-Methyl-1-p-tolylethylamine
Irritant
1-Methyl-1-p-tolylethylamine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1-Methyl-1-p-tolylethylamine
Supplier:
Apollo Scientific
About this item
CAS Number:
6526-79-0
:
Formula:
C₁₀H₁₅N
MDL Number:
MFCD08669637
:
149.237 g/mol
Melting Pt:
112 °C
:
GHS07
Storage Temperature:
Ambient