Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Aminoindan-1-one
5-Aminoindan-1-one
5-Aminoindan-1-one
Irritant
5-Aminoindan-1-one
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
5-Aminoindan-1-one
Supplier:
Apollo Scientific
About this item
CAS Number:
3470-54-0
:
Formula:
C₉H₉NO
MDL Number:
MFCD00099466
:
147.177 g/mol
Melting Pt:
184…186 °C
Storage Temperature:
Ambient
UN:
2811
ADR:
6.1,III
:
GHS07