
3-Amino-4-bromo-5-methyl-1H-pyrazole hydrobromide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:167683-86-5
:
Formula:C₄H₇Br₂N₃
MDL Number:MFCD00053023
:256.94 g/mol
Melting Pt:178…181 °C
:GHS07
Storage Temperature:Ambient