Order Entry
Germany
ContactUsLinkComponent
1,5-Dimethyl-1H-pyrazole 96%
Flammable
Irritant
1,5-Dimethyl-1H-pyrazole 96%
Catalog # APOSOR2504-25G
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Irritant
1,5-Dimethyl-1H-pyrazole 96%
Catalog # APOSOR2504-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :96.1300 g/mol
Storage Temperature:Ambient
UN:1993
ADR:3,III
 :GHS02
 :GHS07
CAS Number:694-31-5
Formula:C₅H₈N₂
MDL Number:MFCD01646129