

Thioanisole 99%
Catalog # APOSOR2490-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:124.21 g/mol
Boiling Pt:188 °C
Melting Pt:–15 °C
Density:1.057 g/cm³ (25 °C)
Flash Pt:57 °C
Storage Temperature:Ambient
EINECS:202-878-2
UN:1993
ADR:3,III
:GHS02
:GHS07
CAS Number:100-68-5
Formula:C₇H₈S
MDL Number:MFCD00008559