Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Quinazoline
Quinazoline 98%
Quinazoline 98%
:
Apollo Scientific
Quinazoline 98%
:
Apollo Scientific
:
253-82-7
:
:
C₈H₆N₂
:
MFCD00006712
:
130.15 g/mol
:
243 °C
:
46…48 °C
:
106 °C
:
Ambient
:
205-965-3
:
GHS07