
Ethyl-5-amino-1-methyl-1H-pyrazole-4-carboxylate
Catalog # APOSOR24192-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:169.18 g/mol
Melting Pt:96…100 °C
EINECS:31037-02-2
:GHS07
Storage Temperature:Ambient
Formula:C₇H₁₁N₃O₂
MDL Number:MFCD00051652