
Ethyl-4-(chlorosulphonyl)-2,5-dimethyl-3-furoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:306936-32-3
:
Formula:C₉H₁₁ClO₅S
MDL Number:MFCD01934807
:266.7 g/mol
Boiling Pt:110…114 °C/0.05mmHg
Storage Temperature:Ambient
UN:3265
ADR:8,III
:GHS05