Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl-4-methyl-1,2,3-thiadiazole-5-carboxylate
Ethyl-4-methyl-1,2,3-thiadiazole-5-carboxylate
Ethyl-4-methyl-1,2,3-thiadiazole-5-carboxylate
Ethyl-4-methyl-1,2,3-thiadiazole-5-carboxylate
APOSOR23649-1G
:
Apollo Scientific
:
Ethyl-4-methyl-1,2,3-thiadiazole-5-carboxylate
APOSOR23649-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
172.21 g/mol
:
117…118 °C/15mmHg
:
1.265 g/cm³ (25 °C)
:
GHS07
:
Ambient
:
18212-20-9
:
C₆H₈N₂O₂S
:
MFCD00052549