Order Entry
Germany
Orders LinkContactUsLinkComponent
Ninhydrin 99%

Ninhydrin 99%

Irritant
Supplier:   Apollo Scientific
Ninhydrin 99%
Supplier:  Apollo Scientific
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate
  • CAS Number:485-47-2
  • Formula:C₉H₆O₄
  • MDL Number:MFCD00003791
  • Molecular Weight:178.14 g/mol
  • Melting Pt:250 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:207-618-1

Documents

Product Family Options

Product Information