
Methyl-5-methylisoxazole-3-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:19788-35-3
:
Formula:C₆H₇NO₃
MDL Number:MFCD00015895
:141.13 g/mol
Boiling Pt:125 °C/11mmHg
Melting Pt:91…92 °C
Storage Temperature:Ambient
EINECS:243-311-9
:GHS07