Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
tert-Butyl thiophen-2-ylcarbamate
tert-Butyl thiophen-2-ylcarbamate
tert-Butyl thiophen-2-ylcarbamate
tert-Butyl thiophen-2-ylcarbamate
Supplier:
Apollo Scientific
Certificates
Expand image
tert-Butyl thiophen-2-ylcarbamate
Supplier:
Apollo Scientific
About this item
CAS Number:
56267-50-6
:
Formula:
C₉H₁₃NO₂S
MDL Number:
MFCD02677742
Melting Pt:
151…152 °C
:
199.2700 g/mol
:
GHS07
Storage Temperature:
Refrigerator