0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
tert-Butyl-3-iodoazetidine-1-carboxylate
tert-Butyl-3-iodoazetidine-1-carboxylate 97%
tert-Butyl-3-iodoazetidine-1-carboxylate 97%
:
Apollo Scientific
tert-Butyl-3-iodoazetidine-1-carboxylate 97%
:
Apollo Scientific
:
254454-54-1
:
:
C₈H₁₄INO₂
:
MFCD09752821
:
283.11 g/mol
:
3082
:
9+EHS,III
:
GHS07
:
GHS09
:
Refrigerator