Order Entry
Germany
ContactUsLinkComponent
 
4-Nitro-2-picoline-N-oxide
Catalog # APOSOR22458-5G
Supplier:  Apollo Scientific
CAS Number:  
4-Nitro-2-picoline-N-oxide
Catalog # APOSOR22458-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 2-Methyl-4-nitropyridine-1-oxide, 4-Nitro-2-picoline N-oxide, 2-Methyl-4-nitropyridine-N-oxide
 :
 :154.13 g/mol
Melting Pt:155…159 °C
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:5470-66-6
Formula:C₆H₆N₂O₃
MDL Number:MFCD00014625