Order Entry
Germany
Orders LinkContactUsLinkComponent
5'-Chloro-2'-hydroxypropiophenone

5'-Chloro-2'-hydroxypropiophenone

Irritant
Supplier:   Apollo Scientific
5'-Chloro-2'-hydroxypropiophenone
Supplier:  Apollo Scientific
Certificates
5'-Chloro-2'-hydroxypropiophenone

2 Options Available Below

Product Details & Documents
  • CAS Number:2892-16-2
  • Formula:C₉H₉ClO₂
  • MDL Number:MFCD00218579
  • Melting Pt:35 °C
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 128,00 €
    128,00 €
    1 * 5 g
       128,00 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 507,00 €
    507,00 €
    1 * 25 g
       507,00 €