5'-Bromo-2'-hydroxypropiophenone
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 1-(5-Bromo-2-hydroxyphenyl)propan-1-one
- CAS Number:17764-93-1
- Formula:C₉H₉BrO₂
- MDL Number:MFCD00218577
- Molecular Weight:229.0706 g/mol
- Melting Pt:76…80 °C
- Storage Temperature:Ambient
- UN:2811
- ADR:6.1,III
2 Options Available Below
Product Details & Documents
Synonyms: 1-(5-Bromo-2-hydroxyphenyl)propan-1-one
- CAS Number:17764-93-1
- Formula:C₉H₉BrO₂
- MDL Number:MFCD00218577
- Molecular Weight:229.0706 g/mol
- Melting Pt:76…80 °C
- Storage Temperature:Ambient
- UN:2811
- ADR:6.1,III



