Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Di-tert-butylpyridine
2,6-Di-tert-butylpyridine
2,6-Di-tert-butylpyridine
:
Apollo Scientific
2,6-Di-tert-butylpyridine
:
Apollo Scientific
:
585-48-8
:
:
C₁₃H₂₁N
:
MFCD00006306
:
191.32 g/mol
:
100…101 °C/23mmHg
:
44288 °C
:
0.852 g/cm³ (25 °C)
:
72 °C
:
209-557-6
:
GHS07
:
Ambient