
N'-Hydroxypyridine-4-carboximidamide
Catalog # APOSOR21874-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:137.142 g/mol
Melting Pt:193…195 °C
:GHS07
Storage Temperature:Ambient
CAS Number:1594-57-6
Formula:C₆H₇N₃O
MDL Number:MFCD00125873