Order Entry
Germany
Orders LinkContactUsLinkComponent
2-(Methylthio)nicotinoyl chloride

2-(Methylthio)nicotinoyl chloride

Corrosive
Supplier:   Apollo Scientific
2-(Methylthio)nicotinoyl chloride
Supplier:  Apollo Scientific
Certificates
2-(Methylthio)nicotinoyl chloride

2 Options Available Below

Product Details & Documents
  • CAS Number:97936-43-1
  • Formula:C₇H₆ClNOS
  • MDL Number:MFCD00051678
  • Molecular Weight:187.65 g/mol
  • Melting Pt:92…96 °C
  • Storage Temperature:Ambient
  • UN:3261
  • ADR:8,II
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 242,00 €
    242,00 €
    1 * 1 g
       242,00 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 484,00 €
    484,00 €
    1 * 5 g
       484,00 €