Order Entry
Germany
ContactUsLinkComponent
3-Amino-4-bromo-5-methylisoxazole
3-Amino-4-bromo-5-methylisoxazole
Catalog # APOSOR21353-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Amino-4-bromo-5-methylisoxazole
Catalog # APOSOR21353-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :177.02 g/mol
Storage Temperature:Ambient
CAS Number:5819-40-9
Formula:C₄H₅BrN₂O
MDL Number:MFCD00052553