Ethyl 3-(4-hydroxy-3-nitrophenyl)propanoate
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:183380-83-8
- Formula:C₁₁H₁₃NO₅
- MDL Number:MFCD00089149
- Molecular Weight:239.23 g/mol
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
- CAS Number:183380-83-8
- Formula:C₁₁H₁₃NO₅
- MDL Number:MFCD00089149
- Molecular Weight:239.23 g/mol
- Storage Temperature:Ambient



