
4-Methyl-1,3,4,5-tetrahydro-2H-1,5-benzodiazepin-2-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3967-01-9
:
Formula:C₁₀H₁₂N₂O
MDL Number:MFCD00024148
:176.22 g/mol
Melting Pt:181…184 °C
:GHS07
Storage Temperature:Ambient