Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Bromo-3,5-xylenol
4-Bromo-3,5-xylenol 98%
4-Bromo-3,5-xylenol 98%
Irritant
4-Bromo-3,5-xylenol 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Bromo-3,5-xylenol 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
7463-51-6
:
Formula:
C₈H₉BrO
MDL Number:
MFCD00002315
Melting Pt:
113…117 °C
:
201.06 g/mol
EINECS:
231-255-8
:
GHS07
Storage Temperature:
Ambient