
6,7-Dimethoxy-1-methyl-1,2,3,4-tetrahydroisoquinoline hydrochloride
Catalog # APOSOR200031-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:243.74 g/mol
Melting Pt:189…192 °C
:GHS07
Storage Temperature:Ambient
CAS Number:63283-42-1
Formula:C₁₂H₁₈ClNO₂
MDL Number:MFCD00050647