0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
5-Bromo-2-iodopyridine
5-Bromo-2-iodopyridine
5-Bromo-2-iodopyridine
5-Bromo-2-iodopyridine
Supplier:
Apollo Scientific
Certificates
Expand image
5-Bromo-2-iodopyridine
Supplier:
Apollo Scientific
About this item
CAS Number:
223463-13-6
:
Formula:
C₅H₃BrIN
MDL Number:
MFCD00691506
:
283.89 g/mol
Melting Pt:
113…117 °C
:
GHS07
Storage Temperature:
Ambient