Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Methoxysalicylaldehyde
5-Methoxysalicylaldehyde 98%
5-Methoxysalicylaldehyde 98%
:
Apollo Scientific
5-Methoxysalicylaldehyde 98%
:
Apollo Scientific
:
672-13-9
:
:
C₈H₈O₃
:
MFCD00003332
:
103 °C/2.5mmHg
:
4 °C
:
1.219 g/cm³ (25 °C)
:
113 °C
:
152.15 g/mol
:
GHS07
:
Ambient