Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
9,10-Dichloroanthracene
9,10-Dichloroanthracene 98%
9,10-Dichloroanthracene 98%
Irritant
9,10-Dichloroanthracene 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
9,10-Dichloroanthracene 98%
Supplier:
Apollo Scientific
Certificates
About this item
CAS Number:
605-48-1
:
Formula:
C₁₄H₈Cl₂
MDL Number:
MFCD00001246
:
247.12 g/mol
Melting Pt:
210…215 °C
EINECS:
210-087-9
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
Documentation