Order Entry
Germany
ContactUsLinkComponent
2-Phenyl-2-propanol 98%
2-Phenyl-2-propanol 98%
Catalog # APOSOR18323-25G
Supplier:  Apollo Scientific
CAS Number:  
2-Phenyl-2-propanol 98%
Catalog # APOSOR18323-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :136.19 g/mol
Boiling Pt:215 °C
Melting Pt:32…34 °C
Density:0.973 g/cm³ (25 °C)
Flash Pt:88 °C
Storage Temperature:Ambient
EINECS:210-539-5
 :GHS07
CAS Number:617-94-7
Formula:C₆H₅C(OH)(CH₃)₂
MDL Number:MFCD00004456