
3-Methyl-2-cyclopenten-1-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2758-18-1
:
Formula:C₆H₈O
MDL Number:MFCD00001403
:96.13 g/mol
Boiling Pt:185…186 °C
Melting Pt:3 °C
Density:0.98 g/cm³ (25 °C)
Flash Pt:65 °C
EINECS:220-421-5
:GHS07
Storage Temperature:Ambient