Order Entry
Germany
ContactUsLinkComponent
tert-Butyl-4-(4-aminophenyl)piperazine-1-carboxylate 98%
tert-Butyl-4-(4-aminophenyl)piperazine-1-carboxylate 98%
Catalog # APOSOR17567-25G
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl-4-(4-aminophenyl)piperazine-1-carboxylate 98%
Catalog # APOSOR17567-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :277.36 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:170911-92-9
Formula:C₁₅H₂₃N₃O₂
MDL Number:MFCD04115065