Order Entry
Germany
ContactUsLinkComponent
tert-Butyl-4-(2-aminoethyl)piperazine-1-carboxylate
tert-Butyl-4-(2-aminoethyl)piperazine-1-carboxylate
Catalog # APOSOR1755-25G
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl-4-(2-aminoethyl)piperazine-1-carboxylate
Catalog # APOSOR1755-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :229.32 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:192130-34-0
Formula:C₁₁H₂₃N₃O₂
MDL Number:MFCD02683049