Order Entry
Germany
ContactUsLinkComponent
1-Boc-3-iodoindole 98%
1-Boc-3-iodoindole 98%
Catalog # APOSOR1733-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Boc-3-iodoindole 98%
Catalog # APOSOR1733-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :343.16 g/mol
Storage Temperature:Ambient
CAS Number:192189-07-4
Formula:C₁₃H₁₄INO₂
MDL Number:MFCD05864781